Butyl-4-hydroxybenzoate ≥99%

Supplier: ACROS ORGANICS MS

Warning

Synonyms: ButylparabenButyl parahydroxybenzoate4-Hydroxybenzoic acid butyl ester

AC40357-1000 AC40357-0025 AC40357-5000
200011-496EA 39.03 USD InStock
200011-496 103514-728 200052-418
Butyl-4-hydroxybenzoate ≥99%
Butyl 4-hydroxybenzoate
Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 156°C (3.5mmHg)
Melting Pt: 73 °C
Density: 1.23 g/cm³ (20 °C)
Flash Pt: 181°C
Storage Temperature: Ambient
MDL Number: MFCD00016478
CAS Number: 94-26-8
EINECS: 202-318-7
UN: 0000
Merck Index: 13,01583

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 68°C to 71°C
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR