Boron trifluoride - methanol complex 12% 1.5 M in methanol
|
Danger
|
AC40276-1000
AC40276-5000
AC40276-0010
200057-208EA
76.7
USD
200057-208
103528-526
200057-206
Boron trifluoride - methanol complex 12% 1.5 M in methanol
Boron trifluoride methanol complex
Formula:
CH₄BF₃O MW: 99.85 g/mol Melting Pt: –98 °C Density: 0.87 g/cm³ Flash Pt: 4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00071635 CAS Number: 373-57-9 EINECS: 206-766-4 UN: 3286 ADR: 3,II |
Specification Test Results
Appearance | Colorless to amber Solution |
Titration with NaOH | 10 to 15 % w/v |
Refractive index | 1.5250 to 1.5300 (20 °C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...