Bicyclohexyl 99%

AC40221-1000 AC40221-0250
103514-716EA 108 USD
103514-716 200005-848
Bicyclohexyl 99%
Bicyclohexyl
Formula: C₁₂H₂₂
MW: 166.31 g/mol
Boiling Pt: 227 °C
Melting Pt: 3…4 °C
Density: 0.86 g/cm³
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00003815
CAS Number: 92-51-3
EINECS: 202-161-4

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4780 to 1.4800 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR