Barium diphenylamine-4-sulfonate, pure indicator grade
|
Warning
|
Synonyms:
Diphenylamine-4-sulphonic acid barium salt
, Diphenylamine-4-sulfonic acid barium salt
, N-Phenylsulfanilic acid barium salt
, N-Phenylsulphanilic acid barium salt
AC40168-0050
AC40168-0100
AC40168-0500
200030-470EA
97.6
USD
200030-470
200024-480
102777-174
Barium diphenylamine-4-sulfonate, pure indicator grade
Barium diphenylamine-4-sulphonate
Formula:
C₂₄H₂₀BaN₂O₆S₂ MW: 633.89 g/mol Melting Pt: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007497 CAS Number: 6211-24-1 EINECS: 228-278-0 UN: 1564 ADR: 6.1,III |
Specification Test Results
Appearance | Off-white to grey Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤1 % (105 °C) |
Sensitivity | (as indicator) Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...