Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aniline blue water soluble

Supplier: ACROS ORGANICS MS

Synonyms: Acid Blue 22Soluble Blue 3M, 2RAniline blue WSMarine Blue VDisodium hydrogen aminomethyl[[4-[(sulphonatophenyl)amino]phenyl][4-[(sulphonatophenyl)imino]cyclohexa-2,5-dien-1-ylidene]methyl]benzenesulphonateChina BlueCotton BlueAniline blue disodiumsaltCI 42755Blackley BlueCN DcK-11Water Blue

AC40118-0250
200005-766EA 100.88 USD InStock
200005-766
Aniline blue water soluble
Aniline blue water soluble


Formula: C₃₂H₂₅N₃Na₂O₉S₃
MW: 737.72 g/mol
Storage Temperature: Ambient
CAS Number: 28631-66-5
EINECS: 249-113-9
Order Now
Specification Test Results
Appearance Dark brown Glistening crystalline powder or crystals
Infrared spectrum Conforms
UV Lambda max 595 to 605 nm (in 0.005M HCl)
Absorption Ratio P-15/P+15 0.96 to 1.05
Titration with TiCl3 ≥12.0 (ml 0.1N TiCl3/g)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty