3,4-Dicyanoaniline 97%

Danger

Synonyms: 4-Aminophthalonitrile

AC40102-0100 AC40102-0050 AC40102-0250
200023-592EA 283 USD
200023-592 200052-110 102776-034
3,4-Dicyanoaniline 97%
3,4-Dicyanoaniline
Formula: C₈H₅N₃
MW: 143.15 g/mol
Boiling Pt: 321 °C (1013 hPa)
Melting Pt: 177…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00041741
CAS Number: 56765-79-8
EINECS: 260-370-6
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow to orange Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 177 to 182 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR