5-Chloro-2-pentanone 90%

Warning

Synonyms: 5-Chloropentan-2-one

AC38829-0250 AC38829-1000
103532-904EA 39.5 USD
103532-904 200054-852
5-Chloro-2-pentanone 90%
5-Chloro-2-pentanone
Formula: C₅H₉ClO
MW: 120.58 g/mol
Boiling Pt: 172…175 °C (760 mmHg)
Melting Pt: –48 °C
Density: 1.053 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00001008
CAS Number: 5891-21-4
EINECS: 227-565-8
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Appearance Brown to black Liquid
Infrared spectrum Conforms
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR