1-Bromoperfluorooctane 99%

Warning

Synonyms: Perfluorooctyl bromide1-BromoheptadecafluorooctanePerflubron

AC38679-0500 AC38679-0100
200056-094EA 104 USD
200056-094 103513-828
1-Bromoperfluorooctane 99%
1-Bromoperfluorooctane
Formula: BrF₂C(CF₂)₆CF₃
MW: 498.96 g/mol
Boiling Pt: 142 °C (760 mmHg)
Melting Pt: 6 °C
Density: 1.93 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00042082
CAS Number: 423-55-2
EINECS: 207-028-4
Merck Index: 12,07299

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR