1-(3-Bromopropoxy)-2-nitrobenzene 90+%

Warning

AC38524-0050 AC38524-0250
200053-544EA 54.65 USD
200053-544 200053-542
1-(3-Bromopropoxy)-2-nitrobenzene 90+%
1-(3-Bromopropoxy)-2-nitrobenzene
Formula: C₉H₁₀BrNO₃
Boiling Pt: 131…132 °C (.5 mmHg)
Melting Pt: 36…39 °C
Flash Pt: 150 °C
CAS Number: 104147-69-5

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 36 to 39 °C
GC ≥90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR