α-Chloro-3,4-difluorotoulene 98%

Danger

Synonyms: 3,4-Difluorobenzyl chloride1-(Chloromethyl)-3,4-difluorobenzene4-(Chloromethyl)-1,2-difluorobenzene

AC38442-0050 AC38442-0250
200054-310EA 73.5 USD
200054-310 200054-308
α-Chloro-3,4-difluorotoulene 98%
α-Chloro-3,4-difluorotoulene
Formula: C₇H₅ClF₂
MW: 162.57 g/mol
MDL Number: MFCD00236218
CAS Number: 698-80-6
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4920 to 1.4940 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR