3,4,5-Trifluorobenzaldehyde ≥98%

AC38421-0010
200054-300EA 32.5 USD
200054-300
3,4,5-Trifluorobenzaldehyde ≥98%
3,4,5-Trifluorobenzaldehyde
Formula: C₇H₃F₃O
MW: 160.1 g/mol
Boiling Pt: 176…183 °C (1013 hPa)
Melting Pt: 25 °C
Density: 1.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00083523
CAS Number: 132123-54-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid after melting
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR