4-Chloro-m-toluidine 98+%

Warning

AC38368-0250 AC38368-0050
200054-630EA 55.2 USD
200054-630 200054-628
4-Chloro-m-toluidine 98+%
4-Chloro-m-toluidine
Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 241 °C (1013 hPa)
Melting Pt: 83…84 °C
MDL Number: MFCD00066332
CAS Number: 7149-75-9
UN: 2239
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
Melting point 83 to 84 °C
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR