Gadolinium trichloride, anhydrous 99.9% (trace metals basis)

Warning

AC38356-0250
200055-632EA 449 USD
200055-632
Gadolinium trichloride, anhydrous 99.9% (trace metals basis)
Gadolinium trichloride
Formula: Cl₃Gd
MW: 263.61 g/mol
Melting Pt: 604…614 °C
MDL Number: MFCD00011024
CAS Number: 10138-52-0
EINECS: 233-386-6

Order Now

Specification Test Results

Appearance White to off-white Powder
Titration Complexometric 58.2 to 61.1 % (Gd)
Total trace metal impurities ≤1000 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR