Phosphorus(III) bromide 1 mol/l in dichloromethane, AcroSeal™

Danger

Synonyms: Phosphorus tribromide

AC38206-1000 AC38206-8000
200056-128EA 461.54 USD
200056-128 200056-126
Phosphorus(III) bromide 1 mol/l in dichloromethane, AcroSeal™
Phosphorus (III) bromide
Formula: PBr₃
MW: 270.69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011436
CAS Number: 7789-60-8
EINECS: 232-178-2
UN: 2922

Order Now

Specification Test Results

Appearance Clear colorless to yellow Solution
Titration Argentometric 0.9 to 1.1 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR