6-Chloro-m-toluic acid 98%

Warning

AC38197-0010 AC38197-0050 AC38197-0250
103513-136EA 551.3 USD
103513-136 200054-098 200054-096
6-Chloro-m-toluic acid 98%
6-Chloro-m-toluic acid
Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Melting Pt: 148…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00045798
CAS Number: 6342-60-5

Order Now

Specification Test Results

Appearance Off-white to beige Powder
Infrared spectrum Conforms
Melting point 148 to 151 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR