2-(3-Phenylpropyl)pyridine 98%
|
Warning
|
AC38194-0010
200053-812EA
22.9
USD
200053-812
2-(3-Phenylpropyl)pyridine 98%
2-(3-Phenylpropyl)pyridine
Formula:
C₁₄H₁₅N MW: 197.28 g/mol Boiling Pt: 115 °C (2 mmHg) Flash Pt: 100 °C |
MDL Number:
MFCD00191237 CAS Number: 2110-18-1 EINECS: 218-300-7 |
Specification Test Results
Appearance | Clear yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5590 to 1.5610 (20 °C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...