1-Fluoro-2-ethynylbenzene 97%

Warning

Synonyms: 1-Ethynyl-2-fluorobenzene(2-Fluorophenyl)ethyne(2-Fluorophenyl)acetylene

AC38099-2500 AC38099-0010
200053-756EA 114 USD
200053-756 200053-758
1-Fluoro-2-ethynylbenzene 97%
1-Fluoro-2-ethynylbenzene
Formula: C₈H₅F
MW: 120.13 g/mol
Flash Pt: 37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00799540
CAS Number: 766-49-4
UN: 1993

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5240 to 1.5270 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR