α,3,5-Tribromotoluene 99%

Danger

Synonyms: alpha,3,5-Tribromotoluene3,5-Dibromobenzyl bromide1,3-Dibromo-5-(bromomethyl)benzene

AC37978-0050
200054-324EA 182 USD
200054-324
α,3,5-Tribromotoluene 99%
α,3,5-Tribromotoluene
Formula: C₇H₅Br₃
MW: 328.83 g/mol
Boiling Pt: 173 °C (19 mmHg)
Melting Pt: 93…97 °C
MDL Number: MFCD00052415
CAS Number: 56908-88-4
UN: 2921
ADR: 8,II

Order Now

Specification Test Results

Appearance White to beige to light brown Powder
Infrared spectrum Conforms
Melting point 93 to 97 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR