3-Bromo-4-chloropyridine 98%

Warning

AC37964-0010 AC37964-0050
200054-388EA 324 USD
200054-388 200054-386
3-Bromo-4-chloropyridine 98%
3-Bromo-4-chloropyridine
Formula: C₅H₃BrClN
MDL Number: MFCD00234008
CAS Number: 36953-42-1

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid after melting
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR