2,4-Dichloropyridine 98%

Warning

AC37947-0050 AC37947-0010
200053-944EA 26.8 USD
200053-944 200053-946
2,4-Dichloropyridine 98%
2,4-Dichloropyridine
Formula: C₅H₃Cl₂N
MW: 147.99 g/mol
Melting Pt: –1 °C
Density: 1.37 g/cm³
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00955618
CAS Number: 26452-80-2
EINECS: 247-717-7
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5500 to 1.5540 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR