1-(2-Bromoethoxy)-2-bromobenzene 95%

Warning

AC37922-0250 AC37922-0050
200051-308EA 33.4 USD
200051-308 200051-306
1-(2-Bromoethoxy)-2-bromobenzene 95%
1-(2-Bromoethoxy)-2-bromobenzene
Formula: C₈H₈Br₂O
MW: 279.96 g/mol
Boiling Pt: 160…162 °C (16 mmHg)
Melting Pt: 35…36 °C
CAS Number: 18800-28-7
UN: 3335

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid after melting (May darken on storage)
Infrared spectrum Conforms
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR