4-Ethylbenzaldehyde 98%

Warning

Synonyms: p-Ethylbenzaldehyde

AC37895-0250 AC37895-1000 AC37895-0050
103513-058EA 165 USD
103513-058 200041-984 200029-804
4-Ethylbenzaldehyde 98%
4-Ethylbenzaldehyde
Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 221 °C (760 mmHg)
Density: 0.97 g/cm³
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00006956
CAS Number: 4748-78-1
EINECS: 225-268-8

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5385 to 1.5405 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR