Diethylamine 99.5%, Extra Pure, redistilled

Danger

AC37837-0025 AC37837-0010 AC37837-1000
200055-462EA 151 USD
200055-462 200055-464 103528-100
Diethylamine 99.5%, Extra Pure, redistilled
Diethylamine
Formula: (C₂H₅)₂NH
MW: 73.14 g/mol
Boiling Pt: 55 °C (760 mmHg)
Melting Pt: –50 °C
Density: 0.71 g/cm³
Flash Pt: –23 °C
Storage Temperature: Ambient
MDL Number: MFCD00009032
CAS Number: 109-89-7
EINECS: 203-716-3
UN: 1154
ADR: 3,II
Merck Index: 13,03138
https://us.vwr.com/stibo/web/std.lang.all/76/20/2297620.jpg

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3840 to 1.3860 (20 °C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥99.4 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR