Ethyl isothiocyanate 96%

Danger

AC37686-1000 AC37686-0050 AC37686-0250
200043-496EA 25.1 USD
200043-496 103532-710 200043-494
Ethyl isothiocyanate 96%
Ethyl isothiocyanate
Formula: C₃H₅NS
MW: 87.15 g/mol
Boiling Pt: 130…132 °C (760 mmHg)
Melting Pt: –6 °C
Density: 0.995 g/cm³
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00004820
CAS Number: 542-85-8
EINECS: 208-831-2
UN: 3286
ADR: 3,II
Merck Index: 13,03848

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5100 to 1.5140 (20 °C, 589 nm)
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR