α-Bromo-3-chloro-5-fluorotoluene 97%

Danger

Synonyms: 1-(Bromomethyl)-3-chloro-5-fluorobenzene3-Chloro-5-fluorobenzyl bromide

AC37470-0050 AC37470-0010
200023-236EA 80 USD
200023-236 200051-988
α-Bromo-3-chloro-5-fluorotoluene 97%
α-Bromo-3-chloro-5-fluorotoluene
Formula: C₇H₅BrClF
MW: 223.47 g/mol
Melting Pt: 25…30 °C
MDL Number: MFCD03788472
CAS Number: 493024-39-8
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance White to off-white Crystals or crystalline mass or liquid
Infrared spectrum Conforms
Melting point 25 to 30 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR