2-Ethylhexyl methacrylate 99% stabilized
|
Warning
|
Synonyms:
Methacrylic acid 2-ethylhexyl ester
AC37117-0010
200034-788EA
86.1
USD
200034-788
2-Ethylhexyl methacrylate 99% stabilized
2-Ethylhexyl methacrylate
Formula:
C₁₂H₂₂O₂ Boiling Pt: 120 °C (18 mmHg) Melting Pt: –50 °C Density: 0.884 g/cm³ Flash Pt: 92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009494 CAS Number: 688-84-6 EINECS: 211-708-6 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4370 to 1.4390 (20 °C, 589 nm) |
GC | ≥98.5 % |
Color scale | ≤15 APHA |
Stabilizer | 40 to 60 ppm MEHQ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...