Ethyl benzoylacetate 90%

Synonyms: Ethyl 3-oxo-3-phenylpropanoate

AC37085-1000 AC37085-0050 AC37085-0500 AC37085-5000
200025-332EA 51.4 USD
200025-332 200019-104 200019-102 102776-050
Ethyl benzoylacetate 90%
Ethyl benzoylacetate
Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: < 0 °C
Density: 1.106 g/cm³
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00009196
CAS Number: 94-02-0
EINECS: 202-295-3
Merck Index: 13,03803

Order Now

Specification Test Results

Appearance Clear colorless to yellow to red to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5240 to 1.5330 (20 °C, 589 nm)
HPLC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR