2-Ethoxyphenol 98%

Danger

Synonyms: Pyrocatechol monoethyl ethero-Ethoxyphenol1-Hydroxy-2-ethoxybenzene

AC37003-5000 AC37003-0050 AC37003-0025 AC37003-1000
103463-514EA 316.5 USD
103463-514 200041-022 200051-854 200051-856
2-Ethoxyphenol 98%
2-Ethoxyphenol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 216…217 °C
Melting Pt: 20…25 °C
Density: 1.09 g/cm³
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00002187
CAS Number: 94-71-3
EINECS: 202-358-5

Order Now

Specification Test Results

Appearance Clear colorless to yellow to pink Liquid or solid
Infrared spectrum Conforms
Refractive index 1.5270 to 1.5310 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR