2-tert-Butylaniline 98%

Warning

Synonyms: 2-(tert-Butyl)aniline

AC36987-0250
200022-924EA 132 USD
200022-924
2-tert-Butylaniline 98%
2-tert-Butylaniline
Formula: C₁₀H₁₅N
MW: 149.24 g/mol
Boiling Pt: 123…124 °C (17 mmHg)
Melting Pt: –60 °C
Density: 0.957 g/cm³
Flash Pt: 102 °C
MDL Number: MFCD00130023
CAS Number: 6310-21-0
EINECS: 228-634-5

Order Now

Specification Test Results

Appearance Clear light yellow to red-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5440 to 1.5460 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR