4-Hydroxyphenylboronic acid 97%

Warning

Synonyms: Dihydroxy(4-hydroxyphenyl)borane(4-Hydroxyphenyl)metaboric acid4-Hydroxybenzeneboronic acid

AC36942-0010 AC36942-0050
200010-474EA 134 USD
200010-474 200010-472
4-Hydroxyphenylboronic acid 97%
4-Hydroxyphenylboronic acid
Formula: C₆H₇BO₃
MW: 137.93 g/mol
Melting Pt: 285 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01074628
CAS Number: 71597-85-8

Order Now

Specification Test Results

Appearance Off-white to light brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR