4-Acetylphenylboronic acid 97%

Warning

AC36940-0250 AC36940-0050 AC36940-0010
200023-554EA 66.8 USD
200023-554 103512-876 200017-328
4-Acetylphenylboronic acid 97%
4-Acetylphenylboronic acid
Formula: C₈H₉BO₃
MW: 163.97 g/mol
Melting Pt: 243…247 °C
Storage Temperature: Ambient
MDL Number: MFCD01074667
CAS Number: 149104-90-5

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR