Platinum(II) acetylacetonate 98%

Warning

AC36935-0250
103517-826EA 1828 USD
103517-826
Platinum(II) acetylacetonate 98%
Platinum (II) acetylacetonate
Formula: C₁₀H₁₄O₄Pt
MW: 393.3 g/mol
Melting Pt: 250…252 °C
Storage Temperature: Ambient
MDL Number: MFCD00000028
CAS Number: 15170-57-7
EINECS: 239-223-5

Order Now

Specification Test Results

Appearance Yellow to yellow-green Powder
Infrared spectrum Conforms
Assay ≥97.5 % (ex Pt)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR