2-Amino-5-bromopyrimidine 97%

Warning

Synonyms: 5-Bromopyrimidin-2-ylamine5-Bromopyrimidin-2-amine

AC36858-0050 AC36858-0010
200022-412EA 123 USD
200022-412 200028-480
2-Amino-5-bromopyrimidine 97%
2-Amino-5-bromopyrimidine
Formula: C₄H₄BrN₃
MW: 174 g/mol
Melting Pt: 239…244 °C
Flash Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012341
CAS Number: 7752-82-1

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 230 to 244 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR