N-Methyl-2-pyrrolidone (NMP) 99.5% for HPLC

Danger

Synonyms: N-Methyl-alpha-pyrrolidone1-Methyl-2-pyrrolidoneNMPN-Methyl-2-pyrrolidoneN-Methylpyrrolidone1-Methyl-2-pyrrolidinoneN-MethylpyrrolidinoneN-methylpyrrolidoneN-Methyl-gamma-butyrolactamN-Methyl-2-pyrrolidinoneM-Pyrol

AC36845-1000 AC36845-0010 AC36845-0025
200015-834EA 105.82 USD
200015-834 200008-712 200033-982
N-Methyl-2-pyrrolidone (NMP) 99.5% for HPLC
N-Methyl-2-pyrrolidone (NMP)
Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: –24 °C
Density: 1.03 g/cm³
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 12,06197
https://us.vwr.com/stibo/web/std.lang.all/88/54/2298854.jpg

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.05 % (K.F.)
GC ≥99.5 %
UV at 285 nm A: ≤1.0
UV at 300 nm A: ≤0.50
UV at 320 nm A: ≤0.10
UV at 340 nm A: ≤0.01
UV at 450 nm A: ≤0.01
Color scale ≤20 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR