Ammonia 2 M in methanol, AcroSeal™

Danger

AC36839-8000 AC36839-1000
103527-930EA 216 USD
103527-930 200057-076
Ammonia 2 M in methanol, AcroSeal™
Ammonia
Formula: NH₃
MW: 17.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011418
CAS Number: 7664-41-7
EINECS: 215-647-6
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Water ≤0.3 %
Strength 1.90 to 2.20 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR