2-Iodobenzaldehyde 98%

Warning

AC36817-0010 AC36817-0050
103512-854EA 331 USD
103512-854 200009-438
2-Iodobenzaldehyde 98%
2-Iodobenzaldehyde
Formula: C₇H₅IO
MW: 232.02 g/mol
Boiling Pt: 129 °C (14 mmHg)
Melting Pt: 34…38 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039570
CAS Number: 26260-02-6

Order Now

Specification Test Results

Appearance White to light beige Powder or low melting crystalline mass
Infrared spectrum Conforms
Melting point 34 to 38 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR