2,3-Dichlorophenylboronic acid 97%

Warning

AC36771-0050 AC36771-0010
200021-924EA 70 USD
200021-924 200021-922
2,3-Dichlorophenylboronic acid 97%
2,3-Dichlorophenylboronic acid
Formula: C₆H₅BCl₂O₂
MW: 190.82 g/mol
Melting Pt: 242…244 °C
Storage Temperature: Ambient
MDL Number: MFCD01075703
CAS Number: 151169-74-3

Order Now

Specification Test Results

Appearance White to cream Powder or crystals
Infrared spectrum Conforms
HPLC ≥96.0 %
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR