2,4-Dichlorophenylboronic acid 97%
|
Warning
|
Synonyms:
2,4-Dichlorobenzeneboronic acid
, Dihydroxy(2,4-dichlorophenyl)borane
, (2,4-Dichlorophenyl)metaboric acid
AC36767-0050
AC36767-0010
200021-932EA
87.3
USD
200021-932
200040-370
2,4-Dichlorophenylboronic acid 97%
2,4-Dichlorophenylboronic acid
Formula:
C₆H₅BCl₂O₂ MW: 190.82 g/mol Melting Pt: 240…245 °C |
MDL Number:
MFCD00013930 CAS Number: 68716-47-2 |
Specification Test Results
Appearance | White to light yellow Powder or crystals |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Titration with NaOH | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...