(Bromomethyl)cyclobutane 99%
|
Warning
|
Synonyms:
Cyclobutyl methyl bromide
AC36764-0050
AC36764-1000
AC36764-0250
200021-290EA
99.8
USD
200021-290
200008-184
103527-922
(Bromomethyl)cyclobutane 99%
(Bromomethyl)cyclobutane
Formula:
C₅H₉Br MW: 149.03 g/mol Boiling Pt: 145…148 °C (760 mmHg) Density: 1.339 g/cm³ Flash Pt: 27 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040960 CAS Number: 17247-58-4 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4790 to 1.4830 (20 °C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...