2-Bromo-4-chloroanisole 98%

Warning

Synonyms: 2-Bromo-4-chloro-1-methoxybenzene

AC36656-0050 AC36656-0250
200022-464EA 86.2 USD
200022-464 200022-466
2-Bromo-4-chloroanisole 98%
2-Bromo-4-chloroanisole
Formula: C₇H₆BrClO
MW: 221.48 g/mol
MDL Number: MFCD00079705
CAS Number: 60633-25-2

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5850 to 1.5870 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR