o,o,o-Tritolyl phosphate 96%

Danger

Synonyms: o,o,o-TCPo,o,o-Tricresyl phosphateTri-o-tolyl phosphateTris(2-methylphenyl) phosphate

AC36646-5000 AC36646-0250 AC36646-1000
200033-086EA 137 USD
200033-086 103532-614 200045-034
o,o,o-Tritolyl phosphate 96%
o,o,o-Tritolyl phosphate
Formula: C₂₁H₂₁O₄P
MW: 368.37 g/mol
Boiling Pt: 410 °C (760 mmHg)
Melting Pt: –25 °C
Density: 1.2 g/cm³
Flash Pt: 225 °C
Storage Temperature: Ambient
MDL Number: MFCD00041906
CAS Number: 78-30-8
EINECS: 201-103-5
UN: 2574
ADR: 6.1,II
Merck Index: 12,09893

Order Now

Specification Test Results

Appearance Clear colorless to yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.5580 to 1.5610 (20 °C, 589 nm)
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR