(2,3-Dichlorophenyl)acetic acid 97%

Warning

AC36555-0250 AC36555-0050
103512-778EA 162.62 USD
103512-778 200003-618
(2,3-Dichlorophenyl)acetic acid 97%
(2,3-Dichlorophenyl)acetic acid
Formula: (C₆H₃Cl₂)CH₂COOH
MW: 205.04 g/mol
Melting Pt: 130…135 °C
Storage Temperature: Ambient
MDL Number: MFCD01861393
CAS Number: 10236-60-9

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 130 to 135 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR