2-Methylpyrrolidine 98%

Danger

AC36539-0020 AC36539-0100
103527-892EA 535 USD
103527-892 200005-122
2-Methylpyrrolidine 98%
2-Methylpyrrolidine
Formula: C₅H₁₁N
MW: 85.15 g/mol
Boiling Pt: 98…100 °C
Density: 0.88 g/cm³
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00014491
CAS Number: 765-38-8
EINECS: 212-144-3
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4350 to 1.4370 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR