3-Ethynylaniline 98%

Warning

Synonyms: (3-Aminophenyl)ethyne(3-Aminophenyl)acetylene1-Amino-3-ethynylbenzene1-Ethynyl-3-aminobenzene

AC36472-0050 AC36472-0250
200009-782EA 204 USD
200009-782 200009-784
3-Ethynylaniline 98%
3-Ethynylaniline
Formula: C₈H₇N
MW: 117.15 g/mol
Boiling Pt: 92…93 °C (2 mmHg)
Melting Pt: 27 °C
Density: 1.04 g/cm³
Flash Pt: 59 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014779
CAS Number: 54060-30-9
EINECS: 258-944-6
UN: 1993

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.6140 to 1.6180 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR