Methanol, extra dry over Molecular Sieve 99.8%, AcroSeal™

Danger

Synonyms: Hydroxy methaneMethyl hydroxideMethyl alcoholMethylolCarbinolWood alcohol

AC36439-1000 AC36439-0010 AC36439-5000 AC36439-0025
200003-588EA 47.2 USD
200003-588 103527-862 103527-864 200003-586
Methanol, extra dry over Molecular Sieve 99.8%, AcroSeal™
Methanol
Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.7 °C (760 mmHg)
Melting Pt: –98 °C
Density: 0.791 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://us.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.005 % (Coulometric)
GC ≥99.8 %
Acidity/alkalinity ≤0.0005 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR