3-Amino-2-bromopyridine 97%

Danger

Synonyms: 2-Bromo-3-pyridylamine2-Bromopyridin-3-amine

AC36060-0010
200003-546EA 35.1 USD
200003-546
3-Amino-2-bromopyridine 97%
3-Amino-2-bromopyridine
Formula: C₅H₅BrN₂
MW: 173.01 g/mol
Melting Pt: 74…79 °C
MDL Number: MFCD00234064
CAS Number: 39856-58-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Beige to dark brown Crystalline powder
Infrared spectrum Conforms
Melting point 74 to 79 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR