2-Amino-3-bromopyridine 98%
|
Warning
|
AC36058-0010
200003-542EA
16.3
USD
200003-542
2-Amino-3-bromopyridine 98%
2-Amino-3-bromopyridine
Formula:
C₅H₅BrN₂ MW: 173.01 g/mol Melting Pt: 61…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095187 CAS Number: 13534-99-1 |
Specification Test Results
Appearance | Grey to brown Powder |
Infrared spectrum | Conforms |
Melting point | 61 to 65 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...