2,5-Dichlorobenzaldehyde 98%

Danger

AC35994-0010 AC35994-0050
200034-394EA 28.6 USD
200034-394 200028-308
2,5-Dichlorobenzaldehyde 98%
2,5-Dichlorobenzaldehyde
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Melting Pt: 54…58 °C
MDL Number: MFCD00156140
CAS Number: 6361-23-5
EINECS: 228-832-1
UN: 3261

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 54 to 58 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR