4-(Trifluoromethyl)phenylboronic acid 98%

Warning

Synonyms: (4-(Trifluoromethyl)phenyl)boronic acid4-(Trifluoromethyl)benzeneboronic acid(4-(Trifluoromethyl)phenyl)metaboric acidDihydroxy(4-(trifluoromethyl)phenyl)borane

AC35976-0050 AC35976-5000
200030-008EA 41.1 USD
200030-008 103512-692
4-(Trifluoromethyl)phenylboronic acid 98%
4-(Trifluoromethyl)phenylboronic acid
Formula: C₇H₆BF₃O₂
MW: 189.93 g/mol
Melting Pt: 245…250 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151855
CAS Number: 128796-39-4

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR