3-Pyridineboronic acid 95%

Warning

Synonyms: 3-Pyridinylboronic acidPyridin-3-ylboronic acidPyridine-3-boronic acid

AC35965-2500 AC35965-0050 AC35965-0010
200020-548EA 24.9 USD
200020-548 103512-682 103512-684
3-Pyridineboronic acid 95%
3-Pyridineboronic acid
Formula: C₅H₆BNO₂
MW: 122.92 g/mol
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00674177
CAS Number: 1692-25-7

Order Now

Specification Test Results

Appearance White to yellow to light orange Powder or crystals and/or chunks
Infrared spectrum Conforms
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR